Assay Details
| Gene Symbol: Phc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042623.2 | NP_001036088.1 | 12 - 13 | 2595 | 75 | |
| NM_001271579.1 | NP_001258508.1 | 13 - 14 | 2722 | 75 | ||
| NM_007905.3 | NP_031931.2 | 13 - 14 | 2751 | 75 | ||
| NR_073364.1 | 13 - 14 | 2758 | 75 | |||
| GenBank mRNA | AK132756.1 | - | 12 - 13 | 2553 | 75 | |
| AK167590.1 | - | 12 - 13 | 2584 | 75 | ||
| BC046535.1 | - | 13 - 14 | 2738 | 4935713 | 75 | |
| BC052394.1 | - | 13 - 14 | 3084 | 5719837 | 75 | |
| S73882.1 | - | 13 - 14 | 2713 | 75 | ||
| S73883.1 | - | 13 - 14 | 2720 | 75 | ||
| S73884.1 | - | 12 - 13 | 2961 | 75 | ||
| U63386.1 | - | 12 - 13 | 2679 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Phc1
Genomic Map