Assay Details
| Gene Symbol: Gcsam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159297.1 | NP_001152769.1 | 5 - 6 | 324 | 85 | |
| NM_008099.4 | NP_032125.3 | 4 - 5 | 258 | 85 | ||
| GenBank mRNA | AK036553.1 | - | 4 - 5 | 276 | 85 | |
| AY500830.1 | - | 5 - 6 | 232 | 85 | ||
| BC099585.1 | - | 4 - 5 | 222 | 30289367 | 85 | |
| U13263.1 | - | 4 - 5 | 259 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2062048 | AK036553.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2062048 | NM_001159297.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2062048 | NM_008099.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 92266 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 21114 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 482727 | AK032941.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 482727 | AK076074.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 482727 | BC059063.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 482727 | BC096549.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 482727 | BC096638.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 482727 | NM_175381.5 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 664926 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 98630 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 197872 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 235700 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 156046 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150224 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136077 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2062048 | AK036553.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2062048 | NM_001159297.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2062048 | NM_008099.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30727341 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30727360 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30727344 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30727350 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30727353 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30727337 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30727335 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30727345 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30727346 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30727333 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30727349 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30727359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30727329 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30727354 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30727342 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30727338 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30727351 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30727332 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30727355 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30727340 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30727347 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30727325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30727358 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30727343 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30727356 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150224 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08585852 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136077 | - | - |
| Whole Mouse Genome G4122A | A_52_P205810 | AK036553.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P205810 | NM_001159297.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P205810 | NM_008099.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104523 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150224 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136077 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13050190 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13050198 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13050178 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13050183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30727355 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727351 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727387 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727365 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727353 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727346 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727349 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12648912 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12651570 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727393 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727397 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727363 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727390 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727384 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13050198 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727357 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727394 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727347 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12550438 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727372 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727350 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727340 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727378 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727333 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727391 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727356 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727359 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727380 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727355 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727354 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12651569 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727439 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727329 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12575265 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727362 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727345 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727375 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12998867 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727342 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727332 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12998856 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727344 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727381 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727360 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30727367 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12648915 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10436025 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10436027 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10436031 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10436032 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10436030 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10436033 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10436028 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10436026 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10436029 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420544_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1442582_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10436024 | - | - |
| Mouse Genome 430 2.0 Array | 1442582_at | - | - |
| Mouse Genome 430 2.0 Array | 1420544_at | - | - |
| Mouse Genome 430 2.0 Array | 1420544_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1442582_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14525_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14525_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600005981.mm.1 | NM_001159297.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600005981.mm.1 | NM_001159297.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600005981.mm.1 | NM_008099.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600005981.mm.1 | NM_008099.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600005981.mm.1 | TC1600000679.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600005982.mm.1 | NM_001159297.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600005982.mm.1 | NM_001159297.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600005982.mm.1 | NM_008099.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600005982.mm.1 | NM_008099.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600005982.mm.1 | TC1600000679.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1420544_at | - | - |
| Mouse Expression Set 430 | 1420544_PM_at | - | - |
| Mouse Expression Set 430 | 1442582_at | - | - |
| Mouse Expression Set 430 | 1442582_PM_at | - | - |
| Murine Genome U74v2 Set | 101147_PM_at | - | - |
| Murine Genome U74v2 Set | 101147_at | - | - |
| Murine Genome U74v2 Set | 130182_PM_at | - | - |
| Murine Genome U74v2 Set | 130182_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420544_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420544_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420544_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420544_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1442582_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1442582_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 816033 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4670377 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4670377 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004670377 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1227716 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1227716 | - | - |
| mouse-6 v1.1 (Illumina) | scl49057.5_195-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4670377 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl49057.5_195-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4670377 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2979702 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2979702 | - | - |

Gcsam
Genomic Map