Assay Details
| Gene Symbol: Gnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008136.2 | NP_032162.2 | 6 - 7 | 1027 | 86 | |
| GenBank mRNA | AK078187.1 | - | 6 - 7 | 990 | 86 | |
| AK186524.1 | - | 1 - 2 | 113 | 86 | ||
| BC010298.1 | - | 6 - 7 | 982 | 3155411 | 86 | |
| BC013243.1 | - | 6 - 7 | 1051 | 86 | ||
| BC094391.1 | - | 4 - 5 | 464 | 86 | ||
| BC110397.1 | - | 2 - 3 | 292 | 86 | ||
| BC129969.1 | - | 4 - 5 | 559 | 86 | ||
| BC129970.1 | - | 4 - 5 | 559 | 86 | ||
| X65026.1 | - | 5 - 6 | 669 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gnl1
Genomic Map