Assay Details
| Gene Symbol: Gpam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008149.3 | NP_032175.2 | 17 - 18 | 2133 | 64 | |
| GenBank mRNA | AK137067.1 | - | 19 - 20 | 2414 | 64 | |
| AK173202.1 | - | 17 - 18 | 2158 | 64 | ||
| BC019201.1 | - | 17 - 18 | 2138 | 5041120 | 64 | |
| BC082776.1 | - | 3 - 4 | 435 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpam
Genomic Map