Assay Details
| Gene Symbol: Hadh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008212.4 | NP_032238.2 | 4 - 5 | 639 | 57 | |
| GenBank mRNA | AK075609.1 | - | 4 - 5 | 565 | 57 | |
| AK132260.1 | - | 4 - 5 | 602 | 57 | ||
| AK148486.1 | - | 4 - 5 | 576 | 57 | ||
| AK167024.1 | - | 4 - 5 | 566 | 57 | ||
| AK168238.1 | - | 4 - 5 | 586 | 57 | ||
| AK168877.1 | - | 4 - 5 | 566 | 57 | ||
| AK169238.1 | - | 2 - 3 | 195 | 57 | ||
| AK169261.1 | - | 4 - 5 | 605 | 57 | ||
| AK211835.1 | - | 1 - 2 | 103 | 57 | ||
| BC064712.1 | - | 4 - 5 | 581 | 30303218 | 57 | |
| D29639.1 | - | 4 - 5 | 565 | 57 | ||
More Information
Set Membership: |
|

Hadh
Genomic Map