Assay Details
| Gene Symbol: Itgb1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008403.4 | NP_032429.1 | 6 - 7 | 722 | 73 | |
| GenBank mRNA | AJ001373.1 | - | 6 - 7 | 704 | 73 | |
| AK002786.1 | - | 6 - 7 | 679 | 73 | ||
| AK004172.1 | - | 6 - 7 | 678 | 73 | ||
| AK007635.1 | - | 6 - 7 | 680 | 73 | ||
| AK075577.1 | - | 6 - 7 | 723 | 73 | ||
| AK089937.1 | - | 6 - 7 | 836 | 73 | ||
| AK210343.1 | - | 1 - 2 | 114 | 73 | ||
| BC021151.1 | - | 6 - 7 | 71 | |||
| BC028772.1 | - | 5 - 6 | 558 | 1230640 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
blood vessel endothelial cell proliferation involved in sprouting angiogenesis
transcription, DNA-templated
regulation of transcription, DNA-templated
negative regulation of protein kinase activity
negative regulation of cell adhesion involved in substrate-bound cell migration
cell adhesion
cell-matrix adhesion
Notch signaling pathway
integrin-mediated signaling pathway
positive regulation of cell proliferation
negative regulation of cell proliferation
positive regulation of endothelial cell migration
negative regulation of fibroblast migration
protein transport
cell differentiation
regulation of cell adhesion
biomineral tissue development
negative regulation of protein binding
activation of protein kinase B activity
integrin activation
regulation of cell adhesion mediated by integrin
tube formation
cellular response to vascular endothelial growth factor stimulus
regulation of GTPase activity
receptor clustering
cellular response to fibroblast growth factor stimulus
positive regulation of Notch signaling pathway
positive regulation of transcription from RNA polymerase II promoter
regulation of blood vessel size
myoblast migration
positive regulation of stress fiber assembly
positive regulation of cell division
positive regulation of focal adhesion assembly
negative regulation of focal adhesion assembly
positive regulation of protein kinase B signaling
negative regulation of ERK1 and ERK2 cascade
negative regulation of cell migration involved in sprouting angiogenesis
positive regulation of protein targeting to membrane
negative regulation of protein targeting to membrane
negative regulation of substrate adhesion-dependent cell spreading
regulation of integrin-mediated signaling pathway
GDP-dissociation inhibitor activity
integrin binding
protein binding
protein C-terminus binding
protein transporter activity
kinase binding
protein kinase binding
protein complex binding
angiogenesis
blood vessel endothelial cell proliferation involved in sprouting angiogenesis
transcription, DNA-templated
regulation of transcription, DNA-templated
negative regulation of protein kinase activity
negative regulation of cell adhesion involved in substrate-bound cell migration
cell adhesion
cell-matrix adhesion
Notch signaling pathway
integrin-mediated signaling pathway
positive regulation of cell proliferation
negative regulation of cell proliferation
positive regulation of endothelial cell migration
negative regulation of fibroblast migration
protein transport
cell differentiation
regulation of cell adhesion
biomineral tissue development
negative regulation of protein binding
activation of protein kinase B activity
integrin activation
regulation of cell adhesion mediated by integrin
tube formation
cellular response to vascular endothelial growth factor stimulus
regulation of GTPase activity
receptor clustering
cellular response to fibroblast growth factor stimulus
positive regulation of Notch signaling pathway
positive regulation of transcription from RNA polymerase II promoter
regulation of blood vessel size
myoblast migration
positive regulation of stress fiber assembly
positive regulation of cell division
positive regulation of focal adhesion assembly
negative regulation of focal adhesion assembly
positive regulation of protein kinase B signaling
negative regulation of ERK1 and ERK2 cascade
negative regulation of cell migration involved in sprouting angiogenesis
positive regulation of protein targeting to membrane
negative regulation of protein targeting to membrane
negative regulation of substrate adhesion-dependent cell spreading
regulation of integrin-mediated signaling pathway
GDP-dissociation inhibitor activity
integrin binding
protein binding
protein C-terminus binding
protein transporter activity
kinase binding
protein kinase binding
protein complex binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28279801 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28279804 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28279803 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28279800 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28279802 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P439267 | BC038615.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P439267 | BC080847.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P439267 | BC096022.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P439267 | NM_001004364.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P439267 | NM_001098168.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P439267 | BC038615.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P439267 | BC080847.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P439267 | BC096022.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P439267 | NM_001004364.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P439267 | NM_001098168.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P106363 | AK147929.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P106363 | BC021151.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P106363 | BC028772.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P106363 | AK002786.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P106363 | AK004172.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 664496 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 89324 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 671961 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 872433 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 506795 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 178278 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 751818 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 742905 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 680396 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 53187 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P439267 | BC038615.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P439267 | BC080847.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P439267 | BC096022.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P439267 | NM_001004364.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P439267 | NM_001098168.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P439267 | BC038615.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P439267 | BC080847.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P439267 | BC096022.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P439267 | NM_001004364.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P439267 | NM_001098168.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P106363 | AK147929.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P106363 | BC021151.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P106363 | BC028772.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P106363 | AK002786.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P106363 | AK004172.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28279790 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28279803 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28279791 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28279776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28279796 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28279792 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28279797 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28279788 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28279789 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28279785 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28279774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28279780 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28279782 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28279778 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28279800 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28279784 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28279799 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07556665 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07556651 | - | - |
| Whole Mouse Genome G4122A | A_52_P439267 | BC038615.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P439267 | BC080847.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P439267 | BC096022.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P439267 | NM_001004364.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P439267 | NM_001098168.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P439267 | BC038615.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P439267 | BC080847.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P439267 | BC096022.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P439267 | NM_001004364.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P439267 | NM_001098168.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P456980 | AJ001373.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106363 | AK147929.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106363 | BC021151.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106363 | BC028772.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106363 | AK002786.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106363 | AK004172.1 | mouse_transcriptome |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28279803 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28279804 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28279800 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28279802 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28279801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28279743 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28279799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28279788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28279767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28279770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28279790 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28279793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28279801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28279768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28279773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28279771 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28279803 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28279778 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28279797 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28279777 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28279785 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28279796 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28279789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28279776 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28279784 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28279780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28279782 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28279792 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10399597 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10399604 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10399601 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10399599 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10399595 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10399598 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10399594 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10399602 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10399596 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10399603 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10399600 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422157_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10399593 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10394862 | - | - |
| Mouse Genome 430 2.0 Array | 1422157_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1422157_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16413_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16413_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200010416.mm.1 | NM_008403.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200010416.mm.1 | NM_008403.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200010416.mm.1 | TC1200001489.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200010419.mm.1 | NM_008403.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200010419.mm.1 | NM_008403.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200010419.mm.1 | TC1200001489.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200010417.mm.1 | NM_008403.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200010417.mm.1 | NM_008403.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200010417.mm.1 | TC1200001489.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422157_a_at | - | - |
| Mouse Expression Set 430 | 1422157_PM_a_at | - | - |
| Murine Genome U74v2 Set | 100991_at | - | - |
| Murine Genome U74v2 Set | 100991_PM_at | - | - |
| Murine Genome U74v2 Set | 100990_PM_g_at | - | - |
| Murine Genome U74v2 Set | 100989_PM_at | - | - |
| Murine Genome U74v2 Set | 100990_g_at | - | - |
| Murine Genome U74v2 Set | 100989_at | - | - |
| Murine Genome U74v2 Set | 170272_r_at | - | - |
| Murine Genome U74v2 Set | 170272_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422157_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422157_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422157_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422157_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3830121 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3830121 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003830121 | - | - |
| mouse-6 v1.1 (Illumina) | scl016413.7_44-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3830121 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl016413.7_44-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3830121 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2924243 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2924243 | - | - |

Itgb1bp1
Genomic Map