Assay Details
| Gene Symbol: Itgb2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008405.3 | NP_032431.2 | 1 - 2 | 75 | 92 | |
| GenBank mRNA | AF051367.1 | - | 1 - 2 | 60 | 92 | |
| AK036640.1 | - | 1 - 2 | 93 | |||
| AK137534.1 | - | 1 - 2 | 76 | 92 | ||
| AK158108.1 | - | 1 - 2 | 76 | 92 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF051367.1,AK137534.1,NM_008405.3 transcripts.
Set Membership: |
|

Itgb2l
Genomic Map