Assay Details
| Gene Symbol: Itgb2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008405.3 | NP_032431.2 | 5 - 6 | 582 | 78 | |
| GenBank mRNA | AF051367.1 | - | 5 - 6 | 567 | 78 | |
| AK036640.1 | - | 5 - 6 | 582 | 78 | ||
| AK137534.1 | - | 5 - 6 | 583 | 78 | ||
| AK158108.1 | - | 5 - 6 | 583 | 78 | ||
| BC113142.1 | - | 5 - 6 | 551 | 40047633 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Itgb2l
Genomic Map