Assay Details
| Gene Symbol: Jrk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008415.6 | NP_032441.4 | 2 | 1824 | 77 | |
| GenBank mRNA | AK042652.1 | - | 2 | 1800 | 77 | |
| AK043134.1 | - | 2 | 1803 | 77 | ||
| AK168885.1 | - | 2 | 1825 | 77 | ||
| AK171809.1 | - | 2 | 1783 | 77 | ||
| BC014281.1 | - | 1 | 1368 | 77 | ||
| U35730.2 | - | 1 | 1750 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30339076 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30339072 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30339073 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30339074 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30339078 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P505143 | BC014281.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P505143 | AK168885.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P505143 | AK171809.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P505143 | NM_008415.6 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P505143 | AK042652.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P505143 | BC014281.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P505143 | AK168885.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P505143 | AK171809.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P505143 | NM_008415.6 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P505143 | AK042652.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 973572 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 594834 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 436863 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128183 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P505143 | BC014281.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P505143 | AK168885.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P505143 | AK171809.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P505143 | NM_008415.6 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P505143 | AK042652.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P505143 | BC014281.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P505143 | AK168885.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P505143 | AK171809.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P505143 | NM_008415.6 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P505143 | AK042652.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339081 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339076 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339079 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30339080 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P128183 | - | - |
| Whole Mouse Genome G4122A | A_52_P63697 | NM_008415.6 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P63697 | AK042652.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P505143 | BC014281.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P505143 | AK168885.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P505143 | AK171809.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P505143 | NM_008415.6 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P505143 | AK042652.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P505143 | BC014281.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P505143 | AK168885.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P505143 | AK171809.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P505143 | NM_008415.6 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P505143 | AK042652.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123821 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P128183 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11998327 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12053100 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12053102 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11998314 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11998323 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12053098 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12053105 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12438082 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12438079 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12053099 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11998335 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12049304 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11998337 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30339076 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30339078 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30339074 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30339072 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30339073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30338989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30338978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30338972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30338999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30338975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30338982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30338997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30338990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30338970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30338985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30338967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30338969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30338986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30338987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30338993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30338996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30338980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30339118 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12145091 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339044 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339034 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339005 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339029 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339052 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339065 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339051 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12323104 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339015 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30338995 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12145105 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12323093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339032 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12254789 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339063 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339089 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12438079 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339026 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12254781 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339053 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339021 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339086 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339048 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339075 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339067 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339003 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12053098 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339001 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339008 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339046 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339072 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12254794 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339023 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339056 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339081 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12323096 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30338996 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339030 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339007 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339037 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30338999 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12254793 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12323081 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12053099 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339013 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30339084 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12145078 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10429498 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10429499 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10429497 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418409_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418410_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10429495 | - | - |
| Mouse Genome 430 2.0 Array | 1418410_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1418409_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1418410_at | - | - |
| Mouse Genome 430 2.0 Array | 1418409_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16469_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16469_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa16640_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500012944.mm.1 | NM_008415.6.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500012944.mm.1 | NM_008415.6 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500012944.mm.1 | TC1500001713.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500012945.mm.1 | NM_008415.6.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500012945.mm.1 | NM_008415.6 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500012945.mm.1 | TC1500001713.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500012947.mm.1 | NM_008415.6.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500012947.mm.1 | NM_008415.6 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500012947.mm.1 | TC1500001713.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500012946.mm.1 | NM_008415.6.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500012946.mm.1 | NM_008415.6 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500012946.mm.1 | TC1500001713.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418410_at | - | - |
| Mouse Expression Set 430 | 1418409_at | - | - |
| Mouse Expression Set 430 | 1418409_PM_at | - | - |
| Mouse Expression Set 430 | 1418410_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418409_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418409_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418410_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418410_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418409_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418409_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418410_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418410_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 534942 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3610181 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003610181 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1234831 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1234831 | - | - |
| mouse-6 v1.1 (Illumina) | scl47085.2_51-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3610181 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl47085.2_51-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3610181 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1234831 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1234831 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl47085.2_51-S | - | - |

Jrk
Genomic Map