Assay Details
| Gene Symbol: Kif3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290805.1 | NP_001277734.1 | 1 - 2 | 159 | 90 | |
| NM_001290806.1 | NP_001277735.1 | 1 - 2 | 159 | 90 | ||
| NM_008443.4 | NP_032469.2 | 1 - 2 | 159 | 90 | ||
| GenBank mRNA | AK147078.1 | - | 1 - 2 | 109 | 90 | |
| AK161223.1 | - | 1 - 2 | 150 | 90 | ||
| AK161920.1 | - | 1 - 2 | 133 | 90 | ||
| AK169425.1 | - | 1 - 2 | 133 | 90 | ||
| BC023936.1 | - | 1 - 2 | 109 | 90 | ||
| BC044720.1 | - | 1 - 2 | 120 | 90 | ||
| BC052707.1 | - | 1 - 2 | 125 | 5702415 | 90 | |
| BY237362.1 | - | 1 - 2 | 149 | 90 | ||
| D12645.1 | - | 1 - 2 | 135 | 90 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK147078.1,AK161223.1,AK161920.1,AK169425.1,BC023936.1,BC044720.1,BC052707.1,D12645.1,NM_001290805.1,NM_001290806.1,NM_008443.4 transcripts.
Set Membership: |
|

Kif3a
Genomic Map