Assay Details
| Gene Symbol: Kif3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008444.4 | NP_032470.3 | 4 - 5 | 1787 | 76 | |
| GenBank mRNA | AK083700.1 | - | 4 - 5 | 1775 | 76 | |
| AK122258.1 | - | 4 - 5 | 1748 | 76 | ||
| AK147467.1 | - | 4 - 5 | 1781 | 76 | ||
| AK148200.1 | - | 4 - 5 | 1768 | 76 | ||
| AK161946.1 | - | 2 - 3 | 227 | 76 | ||
| BC139388.1 | - | 3 - 4 | 1706 | 8862410 | 76 | |
| BC139389.1 | - | 3 - 4 | 1706 | 8862411 | 76 | |
| D26077.1 | - | 3 - 4 | 1700 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kif3b
Genomic Map