Assay Details
| Gene Symbol: Kif3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008445.2 | NP_032471.2 | 2 - 3 | 2507 | 62 | |
| GenBank mRNA | AF013116.1 | - | 2 - 3 | 2083 | 62 | |
| AK139894.1 | - | 2 - 3 | 183 | 62 | ||
| AK147572.1 | - | 2 - 3 | 2309 | 62 | ||
| AK220283.1 | - | 3 - 4 | 2232 | 62 | ||
| BC052732.1 | - | 2 - 3 | 1664 | 62 | ||
| BC053053.1 | - | 2 - 3 | 2449 | 62 | ||
| BC062648.1 | - | 2 - 3 | 2182 | 62 | ||
| BC127063.1 | - | 2 - 3 | 1719 | 40109965 | 62 | |
More Information
Set Membership: |
|

Kif3c
Genomic Map