Assay Details
Gene Symbol: Kif4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008446.2 | NP_032472.2 | 28 - 29 | 3781 | 91 | |
GenBank mRNA | AK084837.1 | - | 15 - 16 | 2117 | 91 | |
AK135430.1 | - | 28 - 29 | 3276 | 91 | ||
AK136346.1 | - | 9 - 10 | 1022 | 91 | ||
AK171938.1 | - | 28 - 29 | 3693 | 91 | ||
BC050946.1 | - | 28 - 29 | 3719 | 6822283 | 91 | |
BC054537.1 | - | 28 - 29 | 3703 | 91 | ||
D12646.1 | - | 28 - 29 | 4108 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
