Assay Details
| Gene Symbol: Klc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008451.2 | NP_032477.2 | 13 - 14 | 1891 | 72 | |
| NR_045528.1 | 13 - 14 | 1923 | 72 | |||
| GenBank mRNA | AF055666.1 | - | 13 - 14 | 1718 | 72 | |
| AK141439.1 | - | 13 - 14 | 1816 | 72 | ||
| BC014845.1 | - | 13 - 14 | 1735 | 2599082 | 72 | |
| CX565742.1 | - | 5 - 6 | 491 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Klc2
Genomic Map