Assay Details
| Gene Symbol: Lamb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277928.1 | NP_001264857.1 | 13 - 14 | 1726 | 75 | |
| NM_008484.2 | NP_032510.2 | 13 - 14 | 1765 | 75 | ||
| GenBank mRNA | AK155769.1 | - | 2578 | 75 | ||
| BC008516.1 | - | 13 - 14 | 1726 | 3592888 | 75 | |
| CK023039.1 | - | 447 | 75 | |||
| L20476.1 | - | 5 - 6 | 526 | 75 | ||
| U43298.1 | - | 13 - 14 | 75 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Lamb3
Genomic Map