Assay Details
| Gene Symbol: Lrmp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281980.1 | NP_001268909.1 | 6 - 7 | 535 | 83 | |
| NM_001281981.1 | NP_001268910.1 | 7 - 8 | 576 | 83 | ||
| NM_008511.3 | NP_032537.3 | 6 - 7 | 807 | 83 | ||
| GenBank mRNA | AK040180.1 | - | 6 - 7 | 808 | 83 | |
| AK050646.1 | - | 6 - 7 | 1245 | 83 | ||
| AK079932.1 | - | 6 - 7 | 765 | 83 | ||
| AK153823.1 | - | 6 - 7 | 805 | 83 | ||
| AK153865.1 | - | 6 - 7 | 537 | 83 | ||
| AK160009.1 | - | 6 - 7 | 767 | 83 | ||
| AK166635.1 | - | 4 - 5 | 256 | 83 | ||
| AK168775.1 | - | 4 - 5 | 233 | 83 | ||
| AK169578.1 | - | 7 - 8 | 560 | 83 | ||
| AY753324.1 | - | 5 - 6 | 370 | 83 | ||
| BC052909.1 | - | 6 - 7 | 743 | 30076553 | 83 | |
| U10484.1 | - | 6 - 7 | 513 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrmp
Genomic Map