Assay Details
| Gene Symbol: Lrmp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281980.1 | NP_001268909.1 | 8 - 9 | 670 | 90 | |
| NM_001281981.1 | NP_001268910.1 | 9 - 10 | 711 | 90 | ||
| NM_008511.3 | NP_032537.3 | 8 - 9 | 942 | 90 | ||
| GenBank mRNA | AK040180.1 | - | 8 - 9 | 943 | 90 | |
| AK050646.1 | - | 8 - 9 | 1380 | 90 | ||
| AK079932.1 | - | 8 - 9 | 900 | 90 | ||
| AK153823.1 | - | 8 - 9 | 940 | 90 | ||
| AK153865.1 | - | 8 - 9 | 672 | 90 | ||
| AK160009.1 | - | 8 - 9 | 902 | 90 | ||
| AK166635.1 | - | 6 - 7 | 391 | 90 | ||
| AK168775.1 | - | 6 - 7 | 368 | 90 | ||
| AK169578.1 | - | 9 - 10 | 695 | 90 | ||
| AY753324.1 | - | 7 - 8 | 505 | 90 | ||
| BC052909.1 | - | 8 - 9 | 878 | 30076553 | 90 | |
| U10484.1 | - | 8 - 9 | 648 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrmp
Genomic Map