Assay Details
| Gene Symbol: Lrp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008513.3 | NP_032539.2 | 9 - 10 | 2217 | 75 | |
| GenBank mRNA | AF064984.1 | - | 9 - 10 | 2207 | 75 | |
| AF077847.1 | - | 9 - 10 | 2095 | 75 | ||
| AK147068.1 | - | 9 - 10 | 2210 | 75 | ||
| AK147231.1 | - | 10 - 11 | 2328 | 75 | ||
| AK147330.1 | - | 10 - 11 | 2328 | 75 | ||
| AK220186.1 | - | 9 - 10 | 2170 | 75 | ||
| BC011374.1 | - | 9 - 10 | 2199 | 3154246 | 75 | |
| BC094610.1 | - | 9 - 10 | 2206 | 75 | ||
More Information
Set Membership: |
|

Lrp5
Genomic Map