Assay Details
| Gene Symbol: Lrp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008513.3 | NP_032539.2 | 15 - 16 | 3548 | 85 | |
| GenBank mRNA | AF064984.1 | - | 15 - 16 | 3538 | 85 | |
| AF077847.1 | - | 15 - 16 | 3426 | 85 | ||
| AK147068.1 | - | 15 - 16 | 3541 | 85 | ||
| AK147231.1 | - | 16 - 17 | 3659 | 85 | ||
| AK147330.1 | - | 16 - 17 | 3659 | 85 | ||
| AK220186.1 | - | 15 - 16 | 3501 | 85 | ||
| BC011374.1 | - | 15 - 16 | 3530 | 3154246 | 85 | |
| BC094610.1 | - | 15 - 16 | 3535 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrp5
Genomic Map