Assay Details
| Gene Symbol: Lrp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008513.3 | NP_032539.2 | 16 - 17 | 3759 | 70 | |
| GenBank mRNA | AF064984.1 | - | 16 - 17 | 3749 | 70 | |
| AF077847.1 | - | 16 - 17 | 3637 | 70 | ||
| AK147068.1 | - | 16 - 17 | 3752 | 70 | ||
| AK147231.1 | - | 17 - 18 | 3870 | 70 | ||
| AK147330.1 | - | 17 - 18 | 3870 | 70 | ||
| AK220186.1 | - | 16 - 17 | 3712 | 70 | ||
| BC011374.1 | - | 16 - 17 | 3741 | 3154246 | 70 | |
| BC094610.1 | - | 16 - 17 | 3746 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrp5
Genomic Map