Assay Details
| Gene Symbol: Lrp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008513.3 | NP_032539.2 | 18 - 19 | 4117 | 62 | |
| GenBank mRNA | AF064984.1 | - | 18 - 19 | 4107 | 62 | |
| AF077847.1 | - | 18 - 19 | 3995 | 62 | ||
| AK090382.1 | - | 9 - 10 | 1792 | 62 | ||
| AK147068.1 | - | 18 - 19 | 4110 | 62 | ||
| AK220186.1 | - | 18 - 19 | 4070 | 62 | ||
| BC011374.1 | - | 18 - 19 | 4099 | 3154246 | 62 | |
| BC094610.1 | - | 18 - 19 | 4104 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrp5
Genomic Map