Assay Details
| Gene Symbol: Ints6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008715.2 | NP_032741.2 | 10 - 11 | 1732 | 102 | |
| GenBank mRNA | AK046055.1 | - | 3 - 4 | 347 | 102 | |
| BC059263.1 | - | 10 - 11 | 1732 | 6409580 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ints6
Genomic Map