Assay Details
| Gene Symbol: Taf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009315.3 | NP_033341.1 | 5 - 6 | 536 | 62 | |
| GenBank mRNA | AK153731.1 | - | 584 | 62 | ||
| AK154867.1 | - | 5 - 6 | 63 | |||
| AK185698.1 | - | 4 - 5 | 325 | 62 | ||
| AK189374.1 | - | 2 - 3 | 156 | 62 | ||
| AK213626.1 | - | 2 - 3 | 156 | 62 | ||
| AK214579.1 | - | 2 - 3 | 156 | 62 | ||
| AK216752.1 | - | 2 - 3 | 156 | 62 | ||
| BC058583.1 | - | 5 - 6 | 536 | 6514960 | 62 | |
| D49439.1 | - | 4 - 5 | 469 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Taf6
Genomic Map