Assay Details
| Gene Symbol: Taf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009315.3 | NP_033341.1 | 10 - 11 | 1159 | 74 | |
| GenBank mRNA | AK153731.1 | - | 1207 | 74 | ||
| AK154867.1 | - | 10 - 11 | 1193 | 74 | ||
| BC058583.1 | - | 10 - 11 | 1159 | 6514960 | 74 | |
| D49439.1 | - | 9 - 10 | 1092 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Taf6
Genomic Map