Assay Details
| Gene Symbol: Tcf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | X61385.1 | - | 2 - 3 | 164 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the X61385.1 transcripts.
Set Membership: |
|

Tcf7
Genomic Map