Assay Details
| Gene Symbol: Inmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009349.3 | NP_033375.1 | 2 - 3 | 374 | 79 | |
| GenBank mRNA | AK002281.1 | - | 2 - 3 | 422 | 79 | |
| AK013010.1 | - | 2 - 3 | 411 | 79 | ||
| BC013518.1 | - | 2 - 3 | 400 | 4236077 | 79 | |
| M88694.1 | - | 2 - 3 | 375 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P162162 | NM_009349.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P162162 | AK002281.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P162162 | AK013010.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P162162 | BC013518.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P162162 | M88694.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P162162 | NM_009349.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P162162 | AK002281.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P162162 | AK013010.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P162162 | BC013518.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P162162 | M88694.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 133010 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 113108 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 290686 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 328957 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 107416 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P152287 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P162162 | NM_009349.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P162162 | AK002281.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P162162 | AK013010.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P162162 | BC013518.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P162162 | M88694.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P162162 | NM_009349.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P162162 | AK002281.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P162162 | AK013010.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P162162 | BC013518.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P162162 | M88694.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467752 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467766 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467779 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467787 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467804 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467802 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467796 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467786 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467769 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467805 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467767 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467751 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467785 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467756 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467783 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467791 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467759 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467763 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467765 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467788 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467797 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467784 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467800 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467772 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467755 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467781 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467750 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467775 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467777 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467798 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467757 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467794 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467790 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24467776 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P152287 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01334657 | - | - |
| Whole Mouse Genome G4122A | A_51_P162162 | NM_009349.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P162162 | AK002281.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P162162 | AK013010.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P162162 | BC013518.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P162162 | M88694.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P162162 | NM_009349.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P162162 | AK002281.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P162162 | AK013010.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P162162 | BC013518.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P162162 | M88694.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113626 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P152287 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05678122 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05434001 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05678124 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05434000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24467722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24467739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24467722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05678104 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05110203 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24467744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05161411 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24467784 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05166286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05678119 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05071124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24467778 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05678124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05879320 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24467771 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05125937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24467736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05678107 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05678112 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05166285 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05690197 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24467772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24467791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05678100 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05034476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24467787 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05166294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05678117 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05434000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05658573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24467765 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05678120 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05166293 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05362342 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24467726 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24467730 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05301716 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24467751 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05658568 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05071123 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24467790 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05362341 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24467716 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05166295 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24467773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05247405 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05161421 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05364528 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24467717 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24467800 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24467718 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05166290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05537304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24467796 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05161426 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05708807 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05658572 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24467752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05362340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24467794 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05161419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24467766 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05678114 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10544935 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10544934 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10544933 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418697_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10544932 | - | - |
| Mouse Genome 430 2.0 Array | 1418697_at | - | - |
| Mouse Genome 430 2.0 Array | 1418697_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21743_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21743_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa14250_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600018191.mm.1 | TC0600002326.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600018191.mm.1 | NM_009349.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600018191.mm.1 | NM_009349.3 | Refseq |
| Mouse Expression Set 430 | 1418697_at | - | - |
| Mouse Expression Set 430 | 1418697_PM_at | - | - |
| Murine Genome U74v2 Set | 97402_PM_at | - | - |
| Murine Genome U74v2 Set | 97402_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418697_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418697_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418697_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418697_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 915779 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2320020 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2320020 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002320020 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1231445 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1231445 | - | - |
| mouse-6 v1.1 (Illumina) | scl28971.3_6-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2320020 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl28971.3_6-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2320020 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2803249 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2803249 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1231445 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl28971.3_6-S | - | - |

Inmt
Genomic Map