Assay Details
| Gene Symbol: Prss39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009355.2 | NP_033381.1 | 4 - 5 | 812 | 90 | |
| GenBank mRNA | AB008910.1 | - | 4 - 5 | 812 | 90 | |
| BC049616.1 | - | 4 - 5 | 807 | 6773970 | 90 | |
| BC115665.2 | - | 4 - 5 | 811 | 40103370 | 90 | |
| BC115666.2 | - | 4 - 5 | 811 | 40103373 | 90 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P392594 | AB008910.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P392594 | AB008910.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 74185 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 884176 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 271201 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119593 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P392594 | AB008910.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P392594 | AB008910.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P392594 | AB008910.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P392594 | AB008910.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139125 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139139 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139115 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139132 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139122 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139119 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139134 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139113 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139140 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139137 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139109 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139127 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139138 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139117 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139152 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139111 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139118 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139154 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139123 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20139147 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119593 | - | - |
| Whole Mouse Genome G4122A | A_51_P392594 | AB008910.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P392594 | AB008910.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124291 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119593 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01044763 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01044777 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01044757 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01044755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20139147 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139118 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139097 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139139 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139150 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139183 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139172 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139111 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139122 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139096 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139100 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139127 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139154 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139116 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139109 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139115 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01015876 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139174 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00689928 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139137 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139087 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139113 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139132 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139168 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139173 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139147 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01044776 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139143 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139167 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139123 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00252067 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139134 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139126 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00836609 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00572094 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139119 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01010141 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139141 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00640700 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139128 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139178 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139152 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139102 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00252083 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00572093 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20139138 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10345391 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10345390 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10345389 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10345392 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10345388 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418559_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10345387 | - | - |
| Mouse Genome 430 2.0 Array | 1418559_at | - | - |
| Mouse Genome 430 2.0 Array | 1418559_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21755_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21755_at | - | - |
| Mouse Expression Set 430 | 1418559_at | - | - |
| Mouse Expression Set 430 | 1418559_PM_at | - | - |
| Murine Genome U74v2 Set | 102244_at | - | - |
| Murine Genome U74v2 Set | 102244_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418559_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418559_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418559_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418559_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 310869 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3990053 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003990053 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2770137 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2770137 | - | - |
| mouse-6 v1.1 (Illumina) | scl18089.5.1_98-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3990053 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl18089.5.1_98-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3990053 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2770137 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2770137 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl18089.5.1_98-S | - | - |

Prss39
Genomic Map