Assay Details
| Gene Symbol: Tgfbi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009369.4 | NP_033395.1 | 8 - 9 | 1171 | 74 | |
| GenBank mRNA | AK135536.1 | - | 8 - 9 | 1216 | 74 | |
| AK142323.1 | - | 8 - 9 | 1191 | 74 | ||
| AK150449.1 | - | 8 - 9 | 1189 | 74 | ||
| AK150833.1 | - | 8 - 9 | 1189 | 74 | ||
| AK150969.1 | - | 8 - 9 | 1190 | 74 | ||
| AK151680.1 | - | 8 - 9 | 74 | |||
| AK152365.1 | - | 8 - 9 | 1190 | 74 | ||
| AK152662.1 | - | 8 - 9 | 1189 | 74 | ||
| AK155572.1 | - | 8 - 9 | 1186 | 74 | ||
| AK155828.1 | - | 8 - 9 | 1186 | 74 | ||
| AK159711.1 | - | 8 - 9 | 1185 | 74 | ||
| AK164373.1 | - | 8 - 9 | 1189 | 74 | ||
| AK169912.1 | - | 8 - 9 | 1191 | 74 | ||
| AK170495.1 | - | 8 - 9 | 1186 | 74 | ||
| AK171557.1 | - | 7 - 8 | 1010 | 74 | ||
| BC129900.1 | - | 8 - 9 | 1164 | 40111549 | 74 | |
| BC129901.1 | - | 8 - 9 | 1164 | 40111550 | 74 | |
| L19932.1 | - | 8 - 9 | 1172 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tgfbi
Genomic Map