Assay Details
| Gene Symbol: Ttc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009441.2 | NP_033467.2 | 18 - 19 | 1712 | 90 | |
| GenBank mRNA | AB008516.1 | - | 17 - 18 | 1589 | 90 | |
| AK053765.1 | - | 18 - 19 | 1816 | 90 | ||
| AK084201.1 | - | 8 - 9 | 747 | 90 | ||
| AK088273.1 | - | 17 - 18 | 1659 | 90 | ||
| AK165850.1 | - | 7 - 8 | 620 | 90 | ||
| AK166120.1 | - | 8 - 9 | 652 | 90 | ||
| AK220495.1 | - | 18 - 19 | 1754 | 90 | ||
| BC019173.1 | - | 18 - 19 | 1681 | 90 | ||
| BC037654.1 | - | 17 - 18 | 1614 | 90 | ||
| BC057207.1 | - | 17 - 18 | 1616 | 90 | ||
| BC145489.1 | - | 13 - 14 | 1232 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ttc3
Genomic Map