Assay Details
| Gene Symbol: Ttc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009441.2 | NP_033467.2 | 24 - 25 | 2350 | 106 | |
| GenBank mRNA | AB008516.1 | - | 23 - 24 | 2227 | 106 | |
| AK084201.1 | - | 14 - 15 | 1385 | 106 | ||
| AK088273.1 | - | 23 - 24 | 2297 | 106 | ||
| AK165850.1 | - | 13 - 14 | 1258 | 106 | ||
| AK166120.1 | - | 14 - 15 | 1290 | 106 | ||
| AK166164.1 | - | 15 - 16 | 1439 | 106 | ||
| AK220495.1 | - | 24 - 25 | 2392 | 106 | ||
| BC019173.1 | - | 24 - 25 | 2319 | 106 | ||
| BC037654.1 | - | 23 - 24 | 2250 | 106 | ||
| BC057207.1 | - | 23 - 24 | 2254 | 106 | ||
| BC145489.1 | - | 19 - 20 | 1870 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ttc3
Genomic Map