Assay Details
| Gene Symbol: Ttc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009441.2 | NP_033467.2 | 25 - 26 | 2410 | 102 | |
| GenBank mRNA | AB008516.1 | - | 24 - 25 | 2287 | 102 | |
| AK084201.1 | - | 15 - 16 | 1445 | 102 | ||
| AK088273.1 | - | 24 - 25 | 2357 | 102 | ||
| AK165850.1 | - | 14 - 15 | 1318 | 102 | ||
| AK166120.1 | - | 15 - 16 | 1350 | 102 | ||
| AK166164.1 | - | 16 - 17 | 1499 | 102 | ||
| AK220495.1 | - | 25 - 26 | 2452 | 102 | ||
| BC019173.1 | - | 25 - 26 | 2379 | 102 | ||
| BC037654.1 | - | 24 - 25 | 2310 | 102 | ||
| BC057207.1 | - | 24 - 25 | 2314 | 102 | ||
| BC145489.1 | - | 20 - 21 | 1930 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ttc3
Genomic Map