Assay Details
| Gene Symbol: Vhl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009507.3 | NP_033533.1 | 2 - 3 | 479 | 84 | |
| GenBank mRNA | AK159238.1 | - | 2 - 3 | 500 | 84 | |
| AK161408.1 | - | 2 - 3 | 476 | 84 | ||
| AK171264.1 | - | 2 - 3 | 481 | 84 | ||
| AK171604.1 | - | 2 - 3 | 489 | 84 | ||
| BC052417.1 | - | 2 - 3 | 478 | 6402536 | 84 | |
| S76748.1 | - | 2 - 3 | 479 | 84 | ||
| U12570.1 | - | 2 - 3 | 495 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
angiogenesis
response to hypoxia
type B pancreatic cell differentiation
pancreatic A cell differentiation
regulation of transcription, DNA-templated
NLS-bearing protein import into nucleus
proteasomal protein catabolic process
protein transport
protein ubiquitination
protein catabolic process
neuron differentiation
extracellular matrix organization
regulation of catecholamine metabolic process
blood vessel endothelial cell migration
negative regulation of endothelial cell differentiation
positive regulation of transcription, DNA-templated
protein heterooligomerization
negative regulation of transcription from RNA polymerase II promoter in response to hypoxia
regulation of thymocyte apoptotic process
negative regulation of thymocyte apoptotic process
negative regulation of hypoxia-inducible factor-1alpha signaling pathway
regulation of apoptotic signaling pathway
protein binding
transcription factor binding
enzyme binding
protein complex binding
ubiquitin protein ligase activity
negative regulation of transcription from RNA polymerase II promoter
angiogenesis
response to hypoxia
type B pancreatic cell differentiation
pancreatic A cell differentiation
regulation of transcription, DNA-templated
NLS-bearing protein import into nucleus
proteasomal protein catabolic process
protein transport
protein ubiquitination
protein catabolic process
neuron differentiation
extracellular matrix organization
regulation of catecholamine metabolic process
blood vessel endothelial cell migration
negative regulation of endothelial cell differentiation
positive regulation of transcription, DNA-templated
protein heterooligomerization
negative regulation of transcription from RNA polymerase II promoter in response to hypoxia
regulation of thymocyte apoptotic process
negative regulation of thymocyte apoptotic process
negative regulation of hypoxia-inducible factor-1alpha signaling pathway
regulation of apoptotic signaling pathway
protein binding
transcription factor binding
enzyme binding
protein complex binding
ubiquitin protein ligase activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24760553 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24760552 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24760554 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P106593 | S76748.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P106593 | BC052417.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P106593 | AK159238.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P106593 | AK171264.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P106593 | NM_009507.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 411906 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 84610 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 546312 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 19369 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 501937 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 48508 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 52118 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P126631 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P106593 | S76748.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P106593 | BC052417.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P106593 | AK159238.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P106593 | AK171264.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P106593 | NM_009507.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P670070 | S76748.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760562 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760538 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760553 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760558 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760555 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760535 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760560 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760546 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760540 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760554 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760559 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760563 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760556 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01421498 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P126631 | - | - |
| Whole Mouse Genome G4122A | A_52_P670070 | S76748.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106593 | S76748.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106593 | BC052417.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106593 | AK159238.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106593 | AK171264.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106593 | NM_009507.3 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P126631 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05233726 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05271971 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24760553 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24760552 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24760554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760635 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760596 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760597 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760563 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760591 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760535 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760577 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760528 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760552 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760531 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760561 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760570 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760588 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760575 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05119633 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05401278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760564 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05271972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760540 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05653734 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760560 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760583 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760554 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760567 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760579 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05450033 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760546 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760538 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05249387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760599 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760585 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05795397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760543 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760562 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760594 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760555 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760587 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10540793 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540791 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540792 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540794 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421071_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434708_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10540790 | - | - |
| Mouse Genome 430 2.0 Array | 1434708_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434708_at | - | - |
| Mouse Genome 430 2.0 Array | 1445514_at | - | - |
| Mouse Genome 430 2.0 Array | 1421071_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1421071_at | - | - |
| Mouse Genome 430 2.0 Array | 1445514_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22346_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22346_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600009891.mm.1 | TC0600001257.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600009891.mm.1 | NM_009507.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600009891.mm.1 | NM_009507.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600009890.mm.1 | TC0600001257.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600009890.mm.1 | NM_009507.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600009890.mm.1 | NM_009507.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600009889.mm.1 | TC0600001257.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600009889.mm.1 | NM_009507.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600009889.mm.1 | NM_009507.3 | Refseq |
| Mouse Expression Set 430 | 1421071_PM_at | - | - |
| Mouse Expression Set 430 | 1421071_at | - | - |
| Mouse Expression Set 430 | 1434708_at | - | - |
| Mouse Expression Set 430 | 1434708_PM_at | - | - |
| Murine Genome U74v2 Set | 102676_at | - | - |
| Murine Genome U74v2 Set | 102676_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421071_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421071_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421071_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421071_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434708_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434708_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2900102 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002900102 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2518546 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2518546 | - | - |
| mouse-6 v1.1 (Illumina) | scl29623.5_373-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2900102 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl29623.5_373-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2900102 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2518546 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2518546 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl29623.5_373-S | - | - |

Vhl
Genomic Map