Assay Details
| Gene Symbol: Was | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009515.2 | NP_033541.1 | 2 - 3 | 339 | 82 | |
| GenBank mRNA | U29673.1 | - | 2 - 3 | 300 | 82 | |
| U54788.1 | - | 2 - 3 | 300 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Was
Genomic Map