Assay Details
| Gene Symbol: Zmat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009517.2 | NP_033543.2 | 2 - 3 | 480 | 86 | |
| GenBank mRNA | AF012923.1 | - | 2 - 3 | 434 | 86 | |
| BC138740.1 | - | 1 - 2 | 299 | 8861762 | 86 | |
| BC138742.1 | - | 1 - 2 | 299 | 8861764 | 86 | |
| BQ895699.1 | - | 2 - 3 | 493 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zmat3
Genomic Map