Assay Details
| Gene Symbol: Ywhae | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF483478.1 | - | 5 - 6 | 715 | 62 | |
| AF483479.1 | - | 5 - 6 | 715 | 62 | ||
| AK169812.1 | - | 5 - 6 | 811 | 62 | ||
| AX775991.1 | - | 4 - 5 | 624 | 62 | ||
| AX775993.1 | - | 3 - 4 | 417 | 62 | ||
| D87663.1 | - | 5 - 6 | 743 | 62 | ||
| Z19599.1 | - | 5 - 6 | 798 | 62 | ||
More Information
Set Membership: |
|

Ywhae
Genomic Map