Assay Details
| Gene Symbol: Plagl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | FJ425893.1 | - | 619 | 123 | ||
| X95503.1 | - | 5 - 6 | 281 | 123 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the X95503.1 transcripts.
Set Membership: |
|

Plagl1
Genomic Map