Assay Details
| Gene Symbol: Plagl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009538.2 | NP_033564.2 | 4 - 5 | 390 | 61 | |
| GenBank mRNA | AF147785.1 | - | 1 - 2 | 83 | 61 | |
| AK142052.1 | - | 5 - 6 | 382 | 61 | ||
| AK142210.1 | - | 6 - 7 | 419 | 61 | ||
| AK183830.1 | - | 4 - 5 | 186 | 61 | ||
| BC065150.1 | - | 4 - 5 | 390 | 61 | ||
| BC141284.1 | - | 5 - 6 | 232 | 9055896 | 61 | |
| BC145612.1 | - | 5 - 6 | 232 | 9054178 | 61 | |
| FJ425893.1 | - | 707 | 61 | |||
| X95503.1 | - | 6 - 7 | 369 | 61 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF147785.1,AK142052.1,AK142210.1,BC141284.1,BC145612.1,NM_009538.2,X95503.1 transcripts.
Set Membership: |
|

Plagl1
Genomic Map