Assay Details
| Gene Symbol: Zap70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289765.1 | NP_001276694.1 | 7 - 8 | 1085 | 100 | |
| NM_001289766.1 | NP_001276695.1 | 7 - 8 | 1050 | 100 | ||
| NM_009539.3 | NP_033565.2 | 7 - 8 | 1042 | 100 | ||
| GenBank mRNA | AK039883.1 | - | 7 - 8 | 979 | 100 | |
| AK089056.1 | - | 7 - 8 | 1015 | 100 | ||
| BC029727.1 | - | 7 - 8 | 1000 | 4925739 | 100 | |
| U04379.1 | - | 7 - 8 | 996 | 100 | ||
| U77667.1 | - | 6 - 7 | 888 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zap70
Genomic Map