Assay Details
| Gene Symbol: Zap70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289612.1 | NP_001276541.1 | 1 - 2 | 333 | 65 | |
| NM_001289765.1 | NP_001276694.1 | 8 - 9 | 1280 | 65 | ||
| NM_001289766.1 | NP_001276695.1 | 8 - 9 | 1245 | 65 | ||
| NM_009539.3 | NP_033565.2 | 8 - 9 | 1237 | 65 | ||
| GenBank mRNA | AB083210.1 | - | 1 - 2 | 336 | 65 | |
| AK039883.1 | - | 8 - 9 | 1174 | 65 | ||
| AK089056.1 | - | 8 - 9 | 1208 | 65 | ||
| BC029727.1 | - | 8 - 9 | 1195 | 4925739 | 65 | |
| U04379.1 | - | 8 - 9 | 1191 | 65 | ||
| U77667.1 | - | 7 - 8 | 1083 | 65 | ||
More Information
Set Membership: |
|

Zap70
Genomic Map