Assay Details
| Gene Symbol: Dpp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159543.1 | NP_001153015.1 | 1 - 2 | 448 | 75 | |
| NM_010074.3 | NP_034204.1 | 1 - 2 | 448 | 75 | ||
| GenBank mRNA | AK153939.1 | - | 1 - 2 | 62 | 75 | |
| AK163085.1 | - | 1 - 2 | 453 | 75 | ||
| X58384.1 | - | 1 - 2 | 125 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK153939.1, AK163085.1, NM_001159543.1, NM_010074.3, X58384.1 transcripts.
Set Membership: |
|

Dpp4
Genomic Map