Assay Details
| Gene Symbol: Dpp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159543.1 | NP_001153015.1 | 21 - 22 | 2310 | 72 | |
| NM_010074.3 | NP_034204.1 | 21 - 22 | 2310 | 72 | ||
| GenBank mRNA | AK085370.1 | - | 21 - 22 | 1893 | 72 | |
| AK153939.1 | - | 21 - 22 | 1924 | 72 | ||
| AK163085.1 | - | 21 - 22 | 2315 | 72 | ||
| AK164775.1 | - | 21 - 22 | 1893 | 72 | ||
| X58384.1 | - | 21 - 22 | 1987 | 72 | ||
More Information
Set Membership: |
|

Dpp4
Genomic Map