Assay Details
| Gene Symbol: Prl8a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010088.2 | NP_034218.1 | 1 - 2 | 91 | 90 | |
| GenBank mRNA | AF011385.1 | - | 1 - 2 | 75 | 90 | |
| AK146049.1 | - | 1 - 2 | 92 | 90 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF011385.1,AK146049.1,NM_010088.2 transcripts.
Set Membership: |
|

Prl8a2
Genomic Map