Assay Details
| Gene Symbol: Gbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010260.1 | NP_034390.1 | 8 - 9 | 1535 | 108 | |
| GenBank mRNA | AF077007.1 | - | 8 - 9 | 1535 | 108 | |
| AF109168.1 | - | 8 - 9 | 1488 | 108 | ||
| AJ007970.1 | - | 8 - 9 | 1528 | 108 | ||
| AK150138.1 | - | 8 - 9 | 1471 | 108 | ||
| AK150146.1 | - | 4 - 5 | 837 | 108 | ||
| BC011336.1 | - | 8 - 9 | 1524 | 3494311 | 108 | |
| BC032882.1 | - | 8 - 9 | 1550 | 1230883 | 108 | |
| BC049141.1 | - | 8 - 9 | 1512 | 108 | ||
| CT010249.1 | - | 7 - 8 | 1365 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gbp2
Genomic Map