Assay Details
| Gene Symbol: Gem | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010276.4 | NP_034406.2 | 1 - 2 | 124 | 86 | |
| GenBank mRNA | AK157655.1 | - | 1 - 2 | 126 | 86 | |
| AK158393.1 | - | 1 - 2 | 126 | 86 | ||
| AK168401.1 | - | 1 - 2 | 125 | 86 | ||
| AK171336.1 | - | 1 - 2 | 62 | 86 | ||
| BC029668.1 | - | 1 - 2 | 100 | 5368352 | 86 | |
| BY133754.1 | - | 1 - 2 | 187 | 86 | ||
| U13053.1 | - | 1 - 2 | 115 | 86 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK158393.1,AK168401.1,AK171336.1,BC029668.1,NM_010276.4,U13053.1 transcripts.
Set Membership: |
|

Gem
Genomic Map