Assay Details
| Gene Symbol: Ggh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF051102.1 | - | 6 - 7 | 689 | 121 | |
| AK089996.1 | - | 6 - 7 | 678 | 121 | ||
| BC009809.1 | - | 6 - 7 | 612 | 4223953 | 121 | |
More Information
Set Membership: |
|

Ggh
Genomic Map