Assay Details
| Gene Symbol: Gna11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010301.3 | NP_034431.1 | 3 - 4 | 594 | 80 | |
| GenBank mRNA | AK143685.1 | - | 3 - 4 | 595 | 80 | |
| M55411.1 | - | 3 - 4 | 531 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gna11
Genomic Map