Assay Details
| Gene Symbol: Gna11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010301.3 | NP_034431.1 | 4 - 5 | 728 | 73 | |
| GenBank mRNA | AK143685.1 | - | 4 - 5 | 729 | 73 | |
| BC011169.1 | - | 4 - 5 | 727 | 4206878 | 73 | |
| M55411.1 | - | 4 - 5 | 665 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gna11
Genomic Map