Assay Details
| Gene Symbol: Gnmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010321.1 | NP_034451.1 | 2 - 3 | 345 | 71 | |
| GenBank mRNA | AK007398.1 | - | 2 - 3 | 172 | 71 | |
| BC014283.1 | - | 2 - 3 | 344 | 4210236 | 71 | |
| BC087960.1 | - | 2 - 3 | 345 | 6766468 | 71 | |
| D89664.1 | - | 2 - 3 | 345 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
glycogen metabolic process
regulation of gluconeogenesis
methionine metabolic process
one-carbon metabolic process
methylation
S-adenosylmethionine metabolic process
protein homotetramerization
folic acid binding
methyltransferase activity
glycine binding
transferase activity
glycine N-methyltransferase activity
glycogen metabolic process
regulation of gluconeogenesis
methionine metabolic process
one-carbon metabolic process
methylation
S-adenosylmethionine metabolic process
protein homotetramerization
folic acid binding
methyltransferase activity
glycine binding
transferase activity
glycine N-methyltransferase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31208633 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31208631 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31208630 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419439 | BC014283.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419439 | D89664.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419439 | NM_010321.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419439 | AK007398.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419439 | BC014283.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419439 | D89664.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419439 | NM_010321.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P419439 | AK007398.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 562250 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 76476 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121358 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419439 | BC014283.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419439 | D89664.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419439 | NM_010321.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419439 | AK007398.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419439 | BC014283.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419439 | D89664.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419439 | NM_010321.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P419439 | AK007398.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P419439 | BC014283.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P419439 | D89664.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P419439 | NM_010321.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P419439 | AK007398.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P419439 | BC014283.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P419439 | D89664.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P419439 | NM_010321.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P419439 | AK007398.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208635 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208644 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208641 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208636 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208634 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208639 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208642 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208630 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208655 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208620 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208647 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208651 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208637 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208638 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208626 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208653 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208623 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208661 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208622 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208660 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208658 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208649 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208648 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208656 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208645 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208646 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208618 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208633 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P121358 | - | - |
| Whole Mouse Genome G4122A | A_51_P419439 | BC014283.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419439 | D89664.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419439 | NM_010321.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419439 | AK007398.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419439 | BC014283.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419439 | D89664.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419439 | NM_010321.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P419439 | AK007398.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100233 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P121358 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13286076 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13286082 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13286069 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13286066 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13081018 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31208633 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31208631 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31208630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208553 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13103291 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208653 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208628 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208618 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208656 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208610 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208647 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13452523 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13181141 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208645 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208621 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208648 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208623 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13266376 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208655 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13286071 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208633 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208663 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13286065 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13103299 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208616 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13140915 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208658 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208622 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208634 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208646 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208630 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208665 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13081018 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13286074 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13181142 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208644 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208624 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208660 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208617 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208615 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13140899 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13286076 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208637 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13286081 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208612 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208641 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10451452 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451454 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451456 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451453 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451457 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451455 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417422_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10451451 | - | - |
| Mouse Genome 430 2.0 Array | 1417422_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417422_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14711_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14711_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700020651.mm.1 | NM_010321.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020651.mm.1 | NM_010321.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020651.mm.1 | TC1700002187.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700020650.mm.1 | NM_010321.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020650.mm.1 | NM_010321.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020650.mm.1 | TC1700002187.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417422_PM_at | - | - |
| Mouse Expression Set 430 | 1417422_at | - | - |
| Murine Genome U74v2 Set | 96828_at | - | - |
| Murine Genome U74v2 Set | 96828_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417422_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417422_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417422_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417422_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 428487 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1410022 | - | - |
| MouseWG-6 v2 Expression BeadChip | 106860053 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106860053 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1410022 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001410022 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1221105 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1221105 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1238783 | - | - |
| mouse-6 v1.1 (Illumina) | scl49855.2.7_13-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106860053 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1410022 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl49855.2.7_13-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106860053 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1410022 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2837816 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1221105 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2837816 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1221105 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1238783 | - | - |

Gnmt
Genomic Map