Assay Details
| Gene Symbol: Got1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010324.2 | NP_034454.2 | 4 - 5 | 683 | 66 | |
| GenBank mRNA | AK146445.1 | - | 4 - 5 | 585 | 66 | |
| AK147934.1 | - | 4 - 5 | 588 | 66 | ||
| AK186873.1 | - | 1 - 2 | 43 | 66 | ||
| AK203412.1 | - | 1 - 2 | 43 | 66 | ||
| BC002057.1 | - | 4 - 5 | 571 | 3489628 | 66 | |
| J02623.1 | - | 4 - 5 | 587 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Got1
Genomic Map