Assay Details
| Gene Symbol: Got2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010325.2 | NP_034455.1 | 7 - 8 | 908 | 71 | |
| GenBank mRNA | AK098166.1 | - | 7 - 8 | 934 | 71 | |
| AK136556.1 | - | 7 - 8 | 945 | 71 | ||
| AK147953.1 | - | 7 - 8 | 948 | 71 | ||
| AK149886.1 | - | 7 - 8 | 945 | 71 | ||
| AK149926.1 | - | 7 - 8 | 945 | 71 | ||
| AK150194.1 | - | 6 - 7 | 664 | 71 | ||
| AK152921.1 | - | 7 - 8 | 945 | 71 | ||
| AK155075.1 | - | 7 - 8 | 939 | 71 | ||
| AK167767.1 | - | 7 - 8 | 934 | 71 | ||
| BC089015.1 | - | 7 - 8 | 934 | 30606379 | 71 | |
| BC089341.1 | - | 7 - 8 | 946 | 6832287 | 71 | |
| J02622.1 | - | 7 - 8 | 908 | 71 | ||
| U82470.1 | - | 7 - 8 | 876 | 71 | ||
| Gene Symbol: Gm10874 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK133132.1 | - | 1 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Got2
Genomic Map